missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclohexanone) oxaldihydrazone, 98%

Product Code. p-7080896
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11433344

Brand: Thermo Scientific Alfa Aesar A10628.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 370-81-0 | C14H22N4O2 | 278.356 g/mol

Bis(cyclohexanone) oxaldihydrazone is used in the quantitative spectroscopic determination of copper. It is also essential for the experimental study of mitochondrial metabolism. It is used to study the human demyelinating diseases such as multiple sclerosis and components of neuroinflammation. Further, it acts as ligand and catalyzes Ullmann-type C-N bond forming reactions in the presence of copper salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(cyclohexanone) oxaldihydrazone is used in the quantitative spectroscopic determination of copper. It is also essential for the experimental study of mitochondrial metabolism. It is used to study the human demyelinating diseases such as multiple sclerosis and components of neuroinflammation. Further, it acts as ligand and catalyzes Ullmann-type C-N bond forming reactions in the presence of copper salt.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 370-81-0
Molecular Formula C14H22N4O2
Molecular Weight (g/mol) 278.356
MDL Number MFCD00001659
InChI Key DSRJIHMZAQEUJV-UHFFFAOYSA-N
Synonym cuprizone, cuprizane, biscyclohexanone oxaldihydrazone, ethanedioic acid, bis cyclohexylidenehydrazide, oxalic acid bis cyclohexylidenehydrazide, biscyclohexanone oxalyldihydrazone, bis cyclohexanone oxaldihydrazone, cuprizon, unii-5n16u7e0ao, oxalic acid, bis cyclohexylidenehydrazide
PubChem CID 9723
IUPAC Name N,N'-bis(cyclohexylideneamino)oxamide
SMILES C1CCC(=NNC(=O)C(=O)NN=C2CCCCC2)CC1

Specifications

Melting Point 208°C to 212°C
Quantity 25 g
Beilstein 2388004
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 278.36
Percent Purity 98%
Chemical Name or Material Bis (cyclohexanone) oxaldihydrazone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.