missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclohexanone) oxaldihydrazone, 98%

Product Code. 11443344
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443344 100 g 100g
11433344 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443344 Supplier Thermo Scientific Alfa Aesar Supplier No. A10628.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 370-81-0 | C14H22N4O2 | 278.356 g/mol

Bis(cyclohexanone) oxaldihydrazone is used in the quantitative spectroscopic determination of copper. It is also essential for the experimental study of mitochondrial metabolism. It is used to study the human demyelinating diseases such as multiple sclerosis and components of neuroinflammation. Further, it acts as ligand and catalyzes Ullmann-type C-N bond forming reactions in the presence of copper salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(cyclohexanone) oxaldihydrazone is used in the quantitative spectroscopic determination of copper. It is also essential for the experimental study of mitochondrial metabolism. It is used to study the human demyelinating diseases such as multiple sclerosis and components of neuroinflammation. Further, it acts as ligand and catalyzes Ullmann-type C-N bond forming reactions in the presence of copper salt.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 370-81-0
Molecular Formula C14H22N4O2
Molecular Weight (g/mol) 278.356
MDL Number MFCD00001659
InChI Key DSRJIHMZAQEUJV-UHFFFAOYSA-N
Synonym cuprizone, cuprizane, biscyclohexanone oxaldihydrazone, ethanedioic acid, bis cyclohexylidenehydrazide, oxalic acid bis cyclohexylidenehydrazide, biscyclohexanone oxalyldihydrazone, bis cyclohexanone oxaldihydrazone, cuprizon, unii-5n16u7e0ao, oxalic acid, bis cyclohexylidenehydrazide
PubChem CID 9723
IUPAC Name N,N'-bis(cyclohexylideneamino)oxamide
SMILES C1CCC(=NNC(=O)C(=O)NN=C2CCCCC2)CC1

Specifications

Melting Point 208°C to 212°C
Quantity 100 g
Beilstein 2388004
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 278.36
Percent Purity 98%
Chemical Name or Material Bis (cyclohexanone) oxaldihydrazone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.