missing translation for 'onlineSavingsMsg'
Learn More

Bis(benzonitrile)palladium(II) Chloride, TRC

Product Code. 30330125
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30330125 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 30330125

Brand: Toronto Research Chemicals TRCB20460810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(benzonitrile)palladium(II) Chloride
CAS 14220-64-5
Molecular Formula C14H10Cl2N2Pd
Quantity 10 mg
Synonym Benzonitrile, Palladium Complex, Bis(benzonitrile)dichloropalladium, Bis(benzonitrile)dichloropalladium(II), Bis(benzonitrile)palladium Dichloride, Bis(benzonitrile)palladium(2+) Chloride, Bis(benzonitrile)palladium(II) Chloride, Bis(phenylnitrile)dichloropalladium, Dibenzonitrilepalladium Dichloride, Dichlorobis(benzonitrile)palladium, Dichlorobis(benzonitrile)palladium(II), Dichlorobis(phenyl cyanide)palladium, NSC 169967, Palladium Chloride Bisbenzonitrile Complex, Palladium Dichloride Bis(benzonitrile)
InChI Formula InChI=1S/2C7H5N.2ClH.Pd/c2*8-6-7-4-2-1-3-5-7;;;/h2*1-5H;2*1H;/q;;;;+2/p-2
SMILES Cl[Pd]Cl.N#Cc1ccccc1.N#Cc2ccccc2
IUPAC Name benzonitrile;dichloropalladium
Molecular Weight (g/mol) 383.57
Formula Weight 381.9256
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.