missing translation for 'onlineSavingsMsg'
Learn More

Bis(benzonitrile)dichloroplatinum(II), Pt 40% min

Product Code. 11326707
Change view
Click to view available options
Quantity:
0.5 g
1 g
5 g
Unit Size:
0.5g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326707 5 g 5g
11336707 0.5 g 0.5g
11316707 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11326707 Supplier Thermo Scientific Alfa Aesar Supplier No. 010004.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 14873-63-3 | C14H10Cl2N2Pt | 472.23 g/mol

Bis(benzonitrile)dichloroplatinum(II) is used as a catalyst for asymmetric hydroformylation reactions, allylation reactions, cyclopropanation reactions, hydrosilylation reactions and carbene insertion into O-H bonds of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14873-63-3
Molecular Formula C14H10Cl2N2Pt
Molecular Weight (g/mol) 472.23
MDL Number MFCD00013125
InChI Key FMBJZWFXNZMJPC-UHFFFAOYSA-L
Synonym dichlorobis benzonitrile platinum ii, bis benzonitrile dichloroplatinum, bis benzonitrile dichloroplatinum ii, cis-bis benzonitrile dichloroplatinum ii, benzonitrile; dichloroplatinum, ptcl2 phcn 2, dichlorobis benzonitrile platinum, dichloro-bis-benzonitrile platinum ii, dichlorobis benzonitrile ;platinum ii ;, bis benzonitrile dichloroplatinate ii
PubChem CID 6093685
IUPAC Name benzonitrile;dichloroplatinum
SMILES Cl[Pt++]Cl.N#CC1=CC=CC=C1.N#CC1=CC=CC=C1

Specifications

Quantity 5 g
UN Number UN3439
Solubility Information Slightly soluble in acetone, chloroform
Formula Weight 472.25
Concentration or Composition (by Analyte or Components) Pt 40% min.
Physical Form Powder
Chemical Name or Material Bis(benzonitrile)dichloroplatinum(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.