missing translation for 'onlineSavingsMsg'
Learn More

Bis(benzonitrile)dichloroplatinum(II), Pt 40% min

Product Code. 11336707
Click to view available options
Quantity:
0.5 g
1 g
5 g
Unit Size:
0.5g
1g
5g
This item is not returnable. View return policy

Product Code. 11336707

Brand: Thermo Scientific Alfa Aesar 010004.02

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14873-63-3 | C14H10Cl2N2Pt | 472.23 g/mol

Bis(benzonitrile)dichloroplatinum(II) is used as a catalyst for asymmetric hydroformylation reactions, allylation reactions, cyclopropanation reactions, hydrosilylation reactions and carbene insertion into O-H bonds of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14873-63-3
Molecular Formula C14H10Cl2N2Pt
Molecular Weight (g/mol) 472.23
MDL Number MFCD00013125
InChI Key FMBJZWFXNZMJPC-UHFFFAOYSA-L
Synonym dichlorobis benzonitrile platinum ii, bis benzonitrile dichloroplatinum, bis benzonitrile dichloroplatinum ii, cis-bis benzonitrile dichloroplatinum ii, benzonitrile; dichloroplatinum, ptcl2 phcn 2, dichlorobis benzonitrile platinum, dichloro-bis-benzonitrile platinum ii, dichlorobis benzonitrile ;platinum ii ;, bis benzonitrile dichloroplatinate ii
PubChem CID 6093685
IUPAC Name benzonitrile;dichloroplatinum
SMILES Cl[Pt++]Cl.N#CC1=CC=CC=C1.N#CC1=CC=CC=C1

Specifications

Quantity 0.5 g
UN Number UN3439
Solubility Information Slightly soluble in acetone, chloroform
Formula Weight 472.25
Concentration or Composition (by Analyte or Components) Pt 40% min.
Physical Form Powder
Chemical Name or Material Bis(benzonitrile)dichloroplatinum(II)
compliance-icons compliance-icons
Product Identifier
  • Bis(benzonitrile)dichloroplatinum(II)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P320-Specific treatment is urgent (see on this label).
  • P361-Take off immediately all contaminated clothing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.