missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis-acrylamide (Electrophoresis), Fisher BioReagents
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
Description
- Bis-acrylamide is a cross-linking agent used in the preparation of polyacrylamide gels for electrophoresis
- RNase, DNase, and protease free
- Specified and tested for use in electrophoresis
Specifications
Absorbance | 0.2 AU max., 0.2 max. (1% solution) at 290nm |
Physical Form | Solid |
Melting Point | 300°C |
Color | White |
pH | 5 |
Quantity | 100 g |
Assay Percent Range | ≥95 % |
DNase | DNase free |
Linear Formula | CH2CHCONHCH2NHCOCHCH2 |
Formula Weight | 154.17 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'