missing translation for 'onlineSavingsMsg'
Learn More

Melford bis-Acrylamide 2% Solution

Product Code. 30017949
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30017949 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30017949 Supplier Melford Supplier No. A11275500.0
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A 2% (w/v) bis-Acrylamide solution.

A 2% (w/v) bis-Acrylamide solution.

Specifications

Chemical Name or Material bis-Acrylamide, 2%
CAS 110-26-9
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.