missing translation for 'onlineSavingsMsg'
Learn More

Bis(acetonitrile)palladium(II) chloride, 99%

Product Code. 10572801 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10572801 500 mg Glass bottle 500mg
10742574 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10572801 Supplier Thermo Scientific Acros Supplier No. 209455000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14592-56-4 | C4H6Cl2N2Pd | 259.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14592-56-4
Molecular Formula C4H6Cl2N2Pd
Molecular Weight (g/mol) 259.42
MDL Number MFCD00013122
InChI Key XGPJSRXOBCGBMN-UHFFFAOYSA-L
IUPAC Name palladium(2+) bis(acetonitrile) dichloride
SMILES CC#N.CC#N.Cl[Pd++]Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 300°C
Color Brown-Rust to Orange
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3CN)2PdCl2
Quantity 500 mg
Fieser 13,33; 15,28; 16,25; 17,30; 01,1015; 03,254; 04,435
Solubility Information Solubility in water: insoluble
Formula Weight 259.4
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Bis(acetonitrile)palladium(II) chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Bis(acetonitrile)palladium(II) chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.