missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(acetonitrile)dichloropalladium(II), TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | Bis(acetonitrile)dichloropalladium(II) |
| CAS | 14592-56-4 |
| Molecular Formula | C4H6Cl2N2Pd |
| Quantity | 2.5 g |
| Synonym | Bis(acetonitrile)palladium Dichloride, Bis(acetonitrile)palladium(II) Dichloride, Diacetonitriledichloropalladium, Dichlorobis(acetonitrile)palladium, Dichlorobis(acetonitrile)palladium(II), Palladium Dichloride Bis(acetonitrile), Bis(Acetonitrile)dichloropalladium |
| InChI Formula | InChI=1S/2C2H3N.2ClH.Pd/c2*1-2-3;;;/h2*1H3;2*1H;/q;;;;+2/p-2 |
| SMILES | [Cl-].[Cl-].[Pd+2].CC#N.CC#N |
| IUPAC Name | acetonitrile;palladium(2+);dichloride |
| Molecular Weight (g/mol) | 259.43 |
| Formula Weight | 257.8943 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction