missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-nitrophenyl) sulfone, 95%, Thermo Scientific™

Product Code. 11479347
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479347 5 g 5g
11489347 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479347 Supplier Thermo Scientific Alfa Aesar Supplier No. B21397.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1156-50-9
Molecular Formula C12H8N2O6S
Molecular Weight (g/mol) 308.264
MDL Number MFCD00010447
InChI Key BVHNGWRPAFKGFP-UHFFFAOYSA-N
Synonym bis 4-nitrophenyl sulfone, 4-nitrophenyl sulfone, 4,4'-dinitrodiphenyl sulfone, bis 4-nitrophenyl sulphone, bis p-nitrophenyl sulfone, 4,4'-dinitrodiphenylsulfone, 1,1'-sulfonylbis 4-nitrobenzene, sulfone, bis p-nitrophenyl, benzene, 1,1'-sulfonylbis 4-nitro-9ci
PubChem CID 70871
IUPAC Name 1-nitro-4-(4-nitrophenyl)sulfonylbenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])S(=O)(=O)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 252°C to 255°C
Odor Odorless
Quantity 5 g
Formula Weight 308.27
Percent Purity 95%
Chemical Name or Material Bis(4-nitrophenyl) sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.