missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-nitrophenyl) carbonate, 98%

Product Code. 11399973
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399973 25 g 25g
11389973 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399973 Supplier Thermo Scientific Alfa Aesar Supplier No. L00290.14
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5070-13-3 | C13H8N2O7 | 304.214 g/mol

Bis(4-nitrophenyl) carbonate is used as a reagent for preparation of symmetrical and unsymmetrical urea and 4-nitrophenyl esters of N-protected amino acids. It acts as a peptide coupling reagent. It finds application as a reagent for the preparation of carbamate linked cytosines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(4-nitrophenyl) carbonate is used as a reagent for preparation of symmetrical and unsymmetrical urea and 4-nitrophenyl esters of N-protected amino acids. It acts as a peptide coupling reagent. It finds application as a reagent for the preparation of carbamate linked cytosines.

Solubility
Soluble in chloroform and tetrahydrofuran.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5070-13-3
Molecular Formula C13H8N2O7
Molecular Weight (g/mol) 304.214
MDL Number MFCD00007322
InChI Key ACBQROXDOHKANW-UHFFFAOYSA-N
Synonym bis 4-nitrophenyl carbonate, bis p-nitrophenyl carbonate, 4,4'-dinitrodiphenyl carbonate, carbonic acid, bis 4-nitrophenyl ester, p,p'-dinitrodiphenylcarbonate, carbonic acid bis 4-nitrophenyl ester, carbonic acid, bis p-nitrophenyl ester, npc bis 4-nitrophenyl carbonate
PubChem CID 78756
IUPAC Name bis(4-nitrophenyl) carbonate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC(=O)OC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 136°C to 139°C
Quantity 25 g
Beilstein 1892897
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform and tetrahydrofuran.
Formula Weight 304.21
Percent Purity 98%
Chemical Name or Material Bis(4-nitrophenyl) carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.