missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-morpholinyl)methane, 98%

Product Code. 15498007
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15498007 5 g 5g
15408017 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498007 Supplier Thermo Scientific Alfa Aesar Supplier No. H51133.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5625-90-1 | C9H18N2O2 | 186.255 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5625-90-1
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD00023369
InChI Key MIFZZKZNMWTHJK-UHFFFAOYSA-N
Synonym dimorpholinomethane, n,n'-dimorpholinomethane, 4,4-methylenedimorpholine, 4,4'-methylenedimorpholine, bismorpholino methane, n,n'-methylenebismorpholine, morpholine, 4,4'-methylenebis, unii-7o79dzw79z, bis morpholino-methan german, bis 4-morpholinyl methane
PubChem CID 21839
IUPAC Name 4-(morpholin-4-ylmethyl)morpholine
SMILES C1COCCN1CN2CCOCC2

Specifications

Boiling Point 122°C to 124°C (12 mmHg)
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 186.25
Percent Purity 98%
Chemical Name or Material Bis(4-morpholinyl)methane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.