missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-methoxybenzyl)amine, 97%, Thermo Scientific Chemicals

Product Code. 15401348
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15401348 1 g 1g
15411348 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15401348 Supplier Thermo Scientific Alfa Aesar Supplier No. H55180.03
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17061-62-0 | Molecular Formula: C16H19NO2 | Molecular Weight: 257.333 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17061-62-0
Molecular Formula C16H19NO2
Molecular Weight (g/mol) 257.333
MDL Number MFCD00277836
InChI Key HBKPDEWGANZHJO-UHFFFAOYSA-N
Synonym bis 4-methoxybenzyl amine, bis-4-methoxy-benzyl-amine, n,n-bis 4-methoxybenzyl amine, bis 4-methoxyphenyl methyl amine, bis-4-methoxybenzyl-amine, benzenemethanamine, 4-methoxy-n-4-methoxyphenyl methyl, bis-4-methoxybenzyl amine, pubchem23909, di 4-methoxybenzyl amine, maybridge1_007781
PubChem CID 714952
IUPAC Name 1-(4-methoxyphenyl)-N-[(4-methoxyphenyl)methyl]methanamine
SMILES COC1=CC=C(C=C1)CNCC2=CC=C(C=C2)OC

Specifications

Quantity 1 g
UN Number UN3259
Formula Weight 257.33
Percent Purity 97%
Chemical Name or Material Bis(4-methoxybenzyl)amine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.