missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-hydroxyphenyl) Sulfone, TRC

Product Code. 30008339
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008339 10 g 10g
30006818 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008339 Supplier Toronto Research Chemicals Supplier No. TRCB44739010G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-09-1
Molecular Formula C12 H10 O4 S
Molecular Weight (g/mol) 250.27
Synonym Phenol, 4,4'-sulfonyldi- (6CI,8CI), 4,4'-Sulfonylbis[phenol], 1,1'-Sulfonylbis[4-hydroxybenzene], 4,4'-Bisphenol S, 4,4'-Dihydroxydiphenyl sulfone, 4,4'-Sulfonyldiphenol, 4-(4-Hydroxyphenylsulfonyl)phenol, 4-Hydroxyphenyl sulfone, BPS 1, BPS-H, BPS-N, BPS-P, BS 3, BS 3 (phenol), BS-PN, Bis(4-hydroxyphenyl) sulfone, Bis(p-hydroxyphenyl) sulfone, Bisphenol S, Bisphenol S sulfonic acid ester, D 8, Diphone A, Diphone C, Diphone D, Dynamar FC 5166, Ex 1B, NSC 683541, NSC 8712, p,p'-Dihydroxydiphenyl sulfone
IUPAC Name 4-(4-hydroxyphenyl)sulfonylphenol
SMILES Oc1ccc(cc1)S(=O)(=O)c2ccc(O)cc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H10O4S/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12/h1-8,13-14H
Formula Weight 250.03
Percent Purity >95%
Chemical Name or Material Bisphenol S
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.