missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-hydroxyphenyl) sulfone, 99%

Product Code. 11413587
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413587 250 g 250g
11403587 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413587 Supplier Thermo Scientific Alfa Aesar Supplier No. A17342.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80-09-1 | C12H10O4S | 250.27 g/mol

It is a reactant used in epoxy reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reactant used in epoxy reactions.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-09-1
Molecular Formula C12H10O4S
Molecular Weight (g/mol) 250.27
MDL Number MFCD00002350
InChI Key VPWNQTHUCYMVMZ-UHFFFAOYSA-N
Synonym 4,4'-sulfonyldiphenol, bisphenol s, bis 4-hydroxyphenyl sulfone, phenol, 4,4'-sulfonylbis, 4,4'-dihydroxydiphenyl sulfone, 4-hydroxyphenyl sulfone, bis p-hydroxyphenyl sulfone, 4,4'-sulfonylbisphenol, diphone c
PubChem CID 6626
ChEBI CHEBI:34372
IUPAC Name 4-(4-hydroxyphenyl)sulfonylphenol
SMILES OC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 245°C to 250°C
Density 1.366
Quantity 250 g
Beilstein 2052954
Solubility Information Insoluble in water.
Formula Weight 250.28
Percent Purity 99%
Chemical Name or Material Bis(4-hydroxyphenyl) sulfone
compliance-icons
Product Identifier
  • Bis(4-hydroxyphenyl) sulfone
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4,4-Sulfonyldiphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.