Learn More
Bis(4-hydroxyphenyl) sulfone, 99%
Description
It is a reactant used in epoxy reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is a reactant used in epoxy reactions.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 245°C to 250°C |
| Density | 1.366 |
| Quantity | 1000 g |
| Beilstein | 2052954 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 250.28 |
| Percent Purity | 99% |
| Chemical Name or Material | Bis(4-hydroxyphenyl) sulfone |
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.