missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-fluorophenyl)methane, 98+%, Thermo Scientific™

Product Code. 11440207
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440207 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11440207 Supplier Thermo Scientific Alfa Aesar Supplier No. B21645.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 457-68-1
Molecular Formula C13H10F2
Molecular Weight (g/mol) 204.22
MDL Number MFCD00000360
InChI Key DXQVFHQUHOFROC-UHFFFAOYSA-N
Synonym bis 4-fluorophenyl methane, 4,4'-difluorodiphenylmethane, 1,1'-methylenebis 4-fluorobenzene, 1-fluoro-4-4-fluorophenyl methyl benzene, benzene, 1,1'-methylenebis 4-fluoro, 4-fluoro-1-4-fluorophenyl methyl benzene, 4,4'-difluorodiphenylamine, bis-4-fluorophenyl methane, 4,4'-difluorodiphenyl methane, 1,1-methylenebis 4-fluorobenzene
PubChem CID 96094
IUPAC Name 1-fluoro-4-[(4-fluorophenyl)methyl]benzene
SMILES C1=CC(=CC=C1CC2=CC=C(C=C2)F)F

Specifications

Melting Point 27°C to 30°C
Density 1.145
Boiling Point 265°C
Flash Point 134°C (273°F)
Refractive Index 1.536
Quantity 25 g
Beilstein 1956387
Solubility Information Insoluble in water.
Formula Weight 204.22
Percent Purity ≥98%
Chemical Name or Material Bis(4-fluorophenyl)methane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.