missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-fluorophenyl) disulfide, 98%

Product Code. 15455337
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455337 1 g 1g
15465337 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455337 Supplier Thermo Scientific Alfa Aesar Supplier No. H32589.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 405-31-2 | C12H8F2S2 | 254.313 g/mol

Bis(4-fluorophenyl) disulfide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(4-fluorophenyl) disulfide is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol and acetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 405-31-2
Molecular Formula C12H8F2S2
Molecular Weight (g/mol) 254.313
MDL Number MFCD00816758
InChI Key SLXZJIKDNHDPKL-UHFFFAOYSA-N
Synonym di-4-fluorophenyl sulfide, bis 4-fluorophenyl disulfide, 1-fluoro-4-4-fluorophenyl disulfanyl benzene, 1,1'-disulfanediylbis 4-fluorobenzene, 4-fluorophenyldisulfide, 4-fluorophenyl disulfide, 4-fluorophenyl disulphide, di-p-fluorophenyl sulfide, bis 4-fluorophenyl persulfide, 1,2-bis 4-fluorophenyl disulfane
PubChem CID 136252
IUPAC Name 1-fluoro-4-[(4-fluorophenyl)disulfanyl]benzene
SMILES C1=CC(=CC=C1F)SSC2=CC=C(C=C2)F

Specifications

Boiling Point 141°C (1 mmHg)
Quantity 1 g
Solubility Information Soluble in methanol and acetone.
Formula Weight 254.31
Percent Purity 98%
Chemical Name or Material Bis(4-fluorophenyl) disulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.