missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-chlorobutyl) ether, 99%

Product Code. 11333946
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11333946 100 g 100g
11313946 5 g 5g
11323946 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11333946 Supplier Thermo Scientific Alfa Aesar Supplier No. L17055.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6334-96-9 | C8H16Cl2O | 199.115 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6334-96-9
Molecular Formula C8H16Cl2O
Molecular Weight (g/mol) 199.115
MDL Number MFCD00019006
InChI Key PVBMXMKIKMJQRK-UHFFFAOYSA-N
Synonym bis 4-chlorobutyl ether, 4,4'-dichlorodibutyl ether, ether, bis 4-chlorobutyl, 4-chlorobutyl ether, di chlorobutyl ether, butane, 1,1'-oxybis 4-chloro, 1,1'-oxydi-4-chlorobutane, 1,1'-oxybis 4-chlorobutane
PubChem CID 22800
IUPAC Name 1-chloro-4-(4-chlorobutoxy)butane
SMILES C(CCCl)COCCCCCl

Specifications

Density 1.081
Boiling Point 129°C to 131°C (10 mmHg)
Linear Formula Cl(CH2)4O(CH2)4)Cl
Refractive Index 1.458
Quantity 100 g
Beilstein 1739861
Formula Weight 199.12
Percent Purity 99%
Chemical Name or Material Bis(4-chlorobutyl) ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.