missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-bromophenyl) ether, 99%

Product Code. 11307208
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11307208

Brand: Thermo Scientific Alfa Aesar 016684.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2050-47-7 | C12H8Br2O | 328.003 g/mol

Bis-(4-bromophenyl)-ether has been used in the preparation of 4-[4-(diethoxyphosphoryl)phenoxy]phenyl-phosphonic acid diethyl ester. It is used for medicine and as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2050-47-7
Molecular Formula C12H8Br2O
Molecular Weight (g/mol) 328.003
MDL Number MFCD00000095
InChI Key YAWIAFUBXXPJMQ-UHFFFAOYSA-N
Synonym bis 4-bromophenyl ether, 4,4'-dibromodiphenyl ether, 4,4'-oxybis bromobenzene, 4-bromophenyl ether, p,p'-dibromodiphenyl ether, usaf do-61, 1-bromo-4-4-bromophenoxy benzene, bis p-bromophenyl ether, ether, bis p-bromophenyl, benzene, 1,1'-oxybis 4-bromo
PubChem CID 16305
IUPAC Name 1-bromo-4-(4-bromophenoxy)benzene
SMILES C1=CC(=CC=C1OC2=CC=C(C=C2)Br)Br

Specifications

Melting Point 56°C to 60°C
Color White
Boiling Point 338°C to 340°C
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 328.01
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Bis(4-bromophenyl) ether

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.