missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-bromophenyl)acetylene, 97%, Thermo Scientific Chemicals

Catalog Number 10579593
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2789-89-1
C14H8Br2
336.03
MFCD00185147
FJQGIJIHOXZMMJ-UHFFFAOYSA-N
1,2-bis 4-bromophenyl ethyne, bis 4-bromophenyl acetylene, 1-bromo-4-2-4-bromophenyl ethynyl benzene, bis-4-bromophenyl acetylene, 1,2-bis 4-bromophenyl acetylene, benzene, 1,1'-1,2-ethynediyl bis 4-bromo, 4,4'-dibromotolan, 4,4'-dibromotolane, bis-p-bromophenyl acetylene, 1 pound not2-bis 4-bromophenyl acetylene
3464242
1-bromo-4-[2-(4-bromophenyl)ethynyl]benzene
BrC1=CC=C(C=C1)C#CC1=CC=C(Br)C=C1
This item is not returnable. View return policy
2789-89-1
C14H8Br2
336.03
MFCD00185147
FJQGIJIHOXZMMJ-UHFFFAOYSA-N
1,2-bis 4-bromophenyl ethyne, bis 4-bromophenyl acetylene, 1-bromo-4-2-4-bromophenyl ethynyl benzene, bis-4-bromophenyl acetylene, 1,2-bis 4-bromophenyl acetylene, benzene, 1,1'-1,2-ethynediyl bis 4-bromo, 4,4'-dibromotolan, 4,4'-dibromotolane, bis-p-bromophenyl acetylene, 1 pound not2-bis 4-bromophenyl acetylene
3464242
1-bromo-4-[2-(4-bromophenyl)ethynyl]benzene
BrC1=CC=C(C=C1)C#CC1=CC=C(Br)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 183°C to 187°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula BrC6H4CCC6H4Br
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 336.02
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-bromophenyl)acetylene
Show More Show Less
compliance-icons
Product Identifier
  • Bis(4-bromophenyl)acetylene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'