missing translation for 'onlineSavingsMsg'
Learn More

Bis[3-(triethoxysilyl)propyl]tetrasulfide, S 22.3% (typical)

Product Code. 11394558
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394558 100 g 100g
11364558 5 g 5g
11374558 25 g 25g
11384558 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11394558 Supplier Thermo Scientific Alfa Aesar Supplier No. 042410.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Coupling/cross-linking agent for precious metals | CAS: 40372-72-3 | C18H42O6S4Si2 | 538.94 g/mol

Bis[3-(triethoxysilyl)propyl]tetrasulfide is used in radial tires, rubber tire treads, carcass, tire walls, solid rubber tires and other rubber products. It acts as a chemical intermediate and as a cross linking agent. Furthermore, it acts as a coupling agent used to chemically modify silica surface. In addition, it serves as a curing agent for good heat aging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40372-72-3
Molecular Formula C18H42O6S4Si2
Molecular Weight (g/mol) 538.94
MDL Number MFCD00053751
InChI Key VTHOKNTVYKTUPI-UHFFFAOYSA-N
Synonym bis 3-triethoxysilyl propyl tetrasulfide, 4,4,15,15-tetraethoxy-3,16-dioxa-8,9,10,11-tetrathia-4,15-disilaoctadecane, unii-j98v193zry, bis 3-triethoxysilyl propyl tetrasulphide, 3,16-dioxa-8,9,10,11-tetrathia-4,15-disilaoctadecane, 4,4,15,15-tetraethoxy, silane coupler kh-858, acmc-1akw5, dsstox_cid_9362
PubChem CID 162012
IUPAC Name triethoxy-[3-(3-triethoxysilylpropyltetrasulfanyl)propyl]silane
SMILES CCO[Si](CCCSSSSCCC[Si](OCC)(OCC)OCC)(OCC)OCC

Specifications

Density 1.074 g/mL
Boiling Point 250°C (decomposition)
Flash Point 91°C (196°F)
Quantity 100 g
Solubility Information Miscible with toluene,primary alcohols,ketones,benzene,chlorinated hydrocarbons,acetonitrile,dimethylformaminde and dimethysulfoxide. Immiscible with water.
Formula Weight 538.94
Concentration or Composition (by Analyte or Components) S 22.3% (typical)
Physical Form Liquid
Chemical Name or Material Bis[3- (triethoxysilyl)propyl]tetrasulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.