missing translation for 'onlineSavingsMsg'
Learn More

Bis(3-aminopropyl)phenylphosphine, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 11355907
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11355907 10 g 10g
11345907 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355907 Supplier Thermo Scientific Alfa Aesar Supplier No. 043675.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6775-01-5
MDL Number MFCD00014834

Specifications

Density 1.03 g/mL
Boiling Point 144°C (1 mmHg)
Packaging ChemSeal™ bottles
Molecular Formula C2H5O3P
Refractive Index 1.5728
Quantity 10 g
UN Number UN3267
Sensitivity Air Sensitive
Synonym phosphonic acid, ethyl, ethylphosphonate, ethanephosphonic acid, phosphate analogue, 6, 4-ethylphosphonic acid, 2-propan-4-ylbenzoic acid, ethylphosphonic acid
InChI Key GATNOFPXSDHULC-UHFFFAOYSA-L
SMILES CCP(=O)(O)O
IUPAC Name ethylphosphonic acid
Molecular Weight (g/mol) 108.03
PubChem CID 204482
Formula Weight 224.29
Physical Form Liquid
Chemical Name or Material Bis (3-aminopropyl)phenylphosphine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.