missing translation for 'onlineSavingsMsg'
Learn More

Bis(3,5,6-trichloro-2-n-pentyloxycarbonylphenyl) oxalate, 95%

Product Code. 15400636
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15400636

Brand: Thermo Scientific Alfa Aesar H53378.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75203-51-9 | C26H24Cl6O8 | 677.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75203-51-9
Molecular Formula C26H24Cl6O8
Molecular Weight (g/mol) 677.17
MDL Number MFCD00012038
InChI Key PURKHUDOTFUVNG-UHFFFAOYSA-N
Synonym cppo, bis 2-carbopentyloxy-3,5,6-trichlorophenyl oxalate, bis 2,3,5-trichloro-6-pentyloxy carbonyl phenyl oxalate, bis 2-carbopentyloxy-3,5,6-trichloro-phenyl oxal, ethanedioic acid, bis 2,3,5-trichloro-6-pentyloxy carbonyl phenyl ester, pentyl 3,4,6-trichloro-2-2-oxo-2-2,3,5-trichloro-6-pentyloxy carbonyl phenoxy acetyl oxy benzoate, bis 2,3,5-trichloro-6-pentoxycarbonylphenyl oxalate, bis 2,3,5-trichloro-6-pentoxycarbonyl-phenyl oxalate
PubChem CID 3018575
IUPAC Name bis(2,3,5-trichloro-6-pentoxycarbonylphenyl) oxalate
SMILES CCCCCOC(=O)C1=C(C(=C(C=C1Cl)Cl)Cl)OC(=O)C(=O)OC2=C(C(=CC(=C2Cl)Cl)Cl)C(=O)OCCCCC

Specifications

Melting Point 76°C to 80°C
Quantity 25 g
Sensitivity Moisture sensitive
Formula Weight 677.19
Percent Purity 95%
Chemical Name or Material Bis(3,5,6-trichloro-2-n-pentyloxycarbonylphenyl) oxalate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.