missing translation for 'onlineSavingsMsg'
Learn More

Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0), 97%

Product Code. 15486886
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15486886 1 g 1g
15496886 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15486886 Supplier Thermo Scientific Alfa Aesar Supplier No. H61964.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 811862-77-8 | C42H44O10Pd | 815.22 g/mol

Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0) used as catalyst in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(3,5,3′,5′-dimethoxydibenzylideneacetone)palladium(0) used as catalyst in pharmaceutical research.

Solubility
Insoluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 811862-77-8
Molecular Formula C42H44O10Pd
Molecular Weight (g/mol) 815.22
MDL Number MFCD07369799
InChI Key RTGAJOPJZJDWAX-UHFFFAOYSA-N
Synonym bis 3,5,3',5'-dimethoxydibenzylideneacetone palladium 0, pd dmdba 2, bis 353'5'-dimethoxydibenzylideneace, bis 3,5,3',5'-dimethoxydibenzylideneacetone palladium
PubChem CID 24777333
SMILES [Pd].COC1=CC(C=CC(=O)C=CC2=CC(OC)=CC(OC)=C2)=CC(OC)=C1.COC1=CC(C=CC(=O)C=CC2=CC(OC)=CC(OC)=C2)=CC(OC)=C1

Specifications

Melting Point 170°C to 178°C
Color Red
Quantity 1 g
Solubility Information Insoluble in water.
IUPAC Name bis(1,5-bis(3,5-dimethoxyphenyl)penta-1,4-dien-3-one) palladium
Formula Weight 815.23
Concentration or Composition (by Analyte or Components) Pd 13%
Physical Form Powder
Chemical Name or Material Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.