missing translation for 'onlineSavingsMsg'
Learn More

Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0), 96%

Product Code. 11936001 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11936001 5 g Glass bottle 5g
11926001 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11936001 Supplier Thermo Scientific Acros Supplier No. 439150050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 811862-77-8 | C42H44O10Pd | 815.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 811862-77-8
Molecular Formula C42H44O10Pd
Molecular Weight (g/mol) 815.22
MDL Number MFCD07369799
InChI Key RTGAJOPJZJDWAX-UHFFFAOYSA-N
Synonym bis 3,5,3',5'-dimethoxydibenzylideneacetone palladium 0, pd dmdba 2, bis 353'5'-dimethoxydibenzylideneace, bis 3,5,3',5'-dimethoxydibenzylideneacetone palladium
PubChem CID 24777333
IUPAC Name 1,5-bis(3,5-dimethoxyphenyl)penta-1,4-dien-3-one;palladium
SMILES [Pd].COC1=CC(C=CC(=O)C=CC2=CC(OC)=CC(OC)=C2)=CC(OC)=C1.COC1=CC(C=CC(=O)C=CC2=CC(OC)=CC(OC)=C2)=CC(OC)=C1

Specifications

Melting Point 170.0°C to 178.0°C
Color Red to Brown
CAS Min % 95.0
CAS Max % 100.0
Physical Form Powder
Quantity 5 g
Assay Percent Range 96%
Formula Weight 815.21
Percent Purity 96%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Bis(3,5,3′,5′-dimethoxydibenzylideneacetone)palladium(0)
Show More Show Less
compliance-icons
Product Identifier
  • Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.