missing translation for 'onlineSavingsMsg'
Learn More

Bis[(2R,5R)-1-(2-[(2R,5R)-2,5-dimethyl-1-phospholanyl]phenyl)-2,5-dimethylphospholane 1-oxide]copper(I) triflate, 97%, Thermo Scientific Chemicals

£54.60 - £204.00

Chemical Identifiers

CAS 874013-62-4
Molecular Formula C37H56CuF3O5P4S
Molecular Weight (g/mol) 857.346
MDL Number MFCD17018797
InChI Key JGVWSKIVCRMXGT-YCIFJGFLSA-M
Synonym bis 1-2r,5r-2,5-dimethylphospholanyl-2-2r,5r-2,5-dimethylphospholanyl-oxide benzene cu i trifluoromethanesulfonate
PubChem CID 11707733
IUPAC Name copper(1+);(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethyl-1$l^{5}-phospholane 1-oxide;trifluoromethanesulfonate
SMILES CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.C(F)(F)(F)S(=O)(=O)[O-].[Cu+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15465615
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63492.MC
100 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15475615
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63492.ME
500 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

874013-62-4
857.346
JGVWSKIVCRMXGT-YCIFJGFLSA-M
11707733
CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.C(F)(F)(F)S(=O)(=O)[O-].[Cu+]
C37H56CuF3O5P4S
MFCD17018797
bis 1-2r,5r-2,5-dimethylphospholanyl-2-2r,5r-2,5-dimethylphospholanyl-oxide benzene cu i trifluoromethanesulfonate
copper(1+);(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethyl-1$l^{5}-phospholane 1-oxide;trifluoromethanesulfonate

Specifications

874013-62-4
C37H56CuF3O5P4S
Air Sensitive
JGVWSKIVCRMXGT-YCIFJGFLSA-M
copper(1+);(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethyl-1$l^{5}-phospholane 1-oxide;trifluoromethanesulfonate
11707733
97%
Bis[(2R,5R)-1-(2-[(2R,5R)-2,5-dimethyl-1-phospholanyl]phenyl)-2,5-dimethylphospholane 1-oxide]copper
Brown to White
MFCD17018797
bis 1-2r,5r-2,5-dimethylphospholanyl-2-2r,5r-2,5-dimethylphospholanyl-oxide benzene cu i trifluoromethanesulfonate
CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C.C(F)(F)(F)S(=O)(=O)[O-].[Cu+]
857.346
857.34
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only