missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-thienyl) Ketone, TRC

Product Code. 30335417
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335417 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335417 Supplier Toronto Research Chemicals Supplier No. TRCB58536510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dithiophen-2-ylmethanone
CAS 704-38-1
Molecular Formula C9 H6 O S2
Quantity 10 g
Synonym Dithiophen-2-ylmethanone, Tiotropium Bromide Monohydrate Imp. F (EP)
InChI Formula InChI=1S/C9H6OS2/c10-9(7-3-1-5-11-7)8-4-2-6-12-8/h1-6H
SMILES O=C(c1cccs1)c2cccs2
IUPAC Name dithiophen-2-ylmethanone
Molecular Weight (g/mol) 194.27
Formula Weight 193.986
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.