missing translation for 'onlineSavingsMsg'
Learn More

Bis-(2-methylallyl)cycloocta-1,5-diene ruthenium(II) complex, 30-32% Ru

Product Code. 10451491 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10451491 250 mg Glass bottle 250mg
10638314 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10451491 Supplier Thermo Scientific Acros Supplier No. 295782500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12289-94-0 | C16H26Ru | 319.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12289-94-0
Molecular Formula C16H26Ru
Molecular Weight (g/mol) 319.45
MDL Number MFCD00216965
InChI Key POYBJJLKGYXKJH-UHFFFAOYSA-N
PubChem CID 91884701
IUPAC Name (5Z)-cycloocta-1,5-diene;2-methanidylprop-1-ene;ruthenium(2+)
SMILES [Ru++].Cc(:c):c.Cc(:c):c.C1C\C=C/CC\C=C/1

Specifications

Melting Point 81°C to 85°C
Color Beige-Brown to Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250 mg
Formula Weight 319.44
Percent Purity 30-32% Ru
Physical Form Crystalline Powder
Chemical Name or Material Bis-(2-methylallyl)cycloocta-1, 5-diene ruthenium(II) complex, 30-32% Ru
compliance-icons
Product Identifier
  • Bis-(2-methylallyl)cycloocta-1,5-diene ruthenium(II) complex
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.