missing translation for 'onlineSavingsMsg'
Learn More

Bis-(2-methylallyl)cycloocta-1,5-diene ruthenium(II) complex, 30-32% Ru

Product Code. 10638314 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10638314

Brand: Thermo Scientific Acros 295780010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12289-94-0 | C16H26Ru | 319.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12289-94-0
Molecular Formula C16H26Ru
Molecular Weight (g/mol) 319.45
MDL Number MFCD00216965
InChI Key POYBJJLKGYXKJH-UHFFFAOYSA-N
PubChem CID 91884701
IUPAC Name (5Z)-cycloocta-1,5-diene;2-methanidylprop-1-ene;ruthenium(2+)
SMILES [Ru++].Cc(:c):c.Cc(:c):c.C1C\C=C/CC\C=C/1

Specifications

Melting Point 81°C to 85°C
Color Beige-Brown to Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 g
Formula Weight 319.44
Percent Purity 30-32% Ru
Physical Form Crystalline Powder
Chemical Name or Material Bis-(2-methylallyl)cycloocta-1, 5-diene ruthenium(II) complex, 30-32% Ru
compliance-icons
Product Identifier
  • Bis-(2-methylallyl)cycloocta-1,5-diene ruthenium(II) complex
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.