missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methylallyl) carbonate, 97%, Thermo Scientific™

Product Code. 13458408 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13458408 25 g Glass bottle 25g
13468408 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13458408 Supplier Thermo Scientific Acros Supplier No. 170580250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 64057-79-0
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00008598
InChI Key SKCUYCAPQSWGBP-UHFFFAOYSA-N
Synonym dimethallyl carbonate, bis 2-methylallyl carbonate, bis methallyl carbonate, bis 2-methyl-2-propenyl carbonate, 2-propen-1-ol, 2-methyl-, carbonate 2:1, 2-methyl-2-propen-1-ol carbonate 2:1, 2-propen-1-ol, 2-methyl-, 1,1'-carbonate, carbonic acid, bis 2-methylallyl ester, bis 2-methylprop-2-en-1-yl carbonate, acmc-1biju
PubChem CID 47083
IUPAC Name bis(2-methylprop-2-enyl) carbonate
SMILES CC(=C)COC(=O)OCC(C)=C

Specifications

Boiling Point 199.0°C to 202.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula [H2C=C(CH3)CH2O]2CO
Refractive Index 1.4371
Quantity 25 g
Formula Weight 170.21
Percent Purity 97%
Chemical Name or Material Bis(2-methylallyl) carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.