missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methoxyethyl) phthalate, 96%, Thermo Scientific Chemicals

Product Code. 11315198
Click to view available options
Quantity:
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It finds it application in cosmetic industry. It is also used as a plasticizer for toys, including inflatable water products, hoppers, play and exercise balls.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-82-8
Molecular Formula C14H18O6
Molecular Weight (g/mol) 282.29
MDL Number MFCD00042842
InChI Key HSUIVCLOAAJSRE-UHFFFAOYSA-N
Synonym bis 2-methoxyethyl phthalate, kesscoflex mcp, 2-methoxyethyl phthalate, dmep, bis methoxyethyl phthalate, di 2-methoxyethyl phthalate, kodaflex dmep, methyl glycol phthalate, dimethoxyethyl phthalate, dimethylglycol phthalate
PubChem CID 8344
IUPAC Name bis(2-methoxyethyl) benzene-1,2-dicarboxylate
SMILES COCCOC(=O)C1=CC=CC=C1C(=O)OCCOC

Specifications

Color Colorless
Density 1.17 g/mL
Boiling Point 340°C
Flash Point 185°C (365°F)
Refractive Index 1.503
Quantity 50 g
Beilstein 2056929
Solubility Information Soluble in water 8500mg/L at 25°C.
Formula Weight 282.29
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Bis (2-methoxyethyl) phthalate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'