missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methoxyethyl) ether, Dr. Ehrenstorfer

Product Code. 30315829
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315829 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315829 Supplier Dr. Ehrenstorfer Supplier No. DREC10653800
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(2-methoxyethyl)ether (Diethylene glycol dimethyl ether)
CAS 111-96-6
Analyte or Component Names Bis(2-methoxyethyl)ether (Diethylene glycol dimethyl ether)
Shipping Condition Room Temperature
Molecular Formula C6 H14 O3
Certifications/Compliance ISO 17034
Synonym Diglyme, 1-Methoxy-2-(2-methoxyethoxy)ethane, Bis(2-methoxyethyl)ether, Diethylene glycol dimethyl ether
InChI Formula InChI=1S/C6H14O3/c1-7-3-5-9-6-4-8-2/h3-6H2,1-2H3
Solution Type Neat
SMILES COCCOCCOC
IUPAC Name 1-methoxy-2-(2-methoxyethoxy)ethane
Industry Type Environmental Testing
Molecular Weight (g/mol) 134.17
Quantity 1 g
Formula Weight 134.0943 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.