missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methoxyethyl) ether, 99%, extra pure

Product Code. 10418860 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10418860 1 L Glass bottle 1 L
10183170 250 mL Glass bottle 250mL
10655172 2.5 L Glass bottle 2.5 L
11641759 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10418860 Supplier Thermo Scientific Acros Supplier No. 125850010
  • ONLINE EXCLUSIVE PRICE
    £101.00
    £97.10 / 1 L
    Save £3.90
    3% Off
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 111-96-6 | C6H14O3 | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-96-6
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
InChI Key SBZXBUIDTXKZTM-UHFFFAOYSA-N
Synonym diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
PubChem CID 8150
ChEBI CHEBI:46784
IUPAC Name 1-methoxy-2-(2-methoxyethoxy)ethane
SMILES COCCOCCOC

Specifications

Melting Point -64.0°C
Density 0.9370g/mL
Boiling Point 162.0°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3OCH2CH2)2O
Refractive Index 1.4070 to 1.4090
Quantity 1 L
Beilstein 01,IV,2393
Fieser 01,255
Merck Index 15,3184
Specific Gravity 0.937
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,ether,hydrocarbon solvents
Viscosity 2 mPa.s (20°C)
Formula Weight 134.18
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Bis(2-methoxyethyl) ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methoxyethyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H226-Flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.