missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methoxyethyl) adipate, 98%, Thermo Scientific™

Product Code. 11478733
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478733 25 g 25g
11488733 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478733 Supplier Thermo Scientific Alfa Aesar Supplier No. A10497.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-00-3
Molecular Formula C12H22O6
Molecular Weight (g/mol) 262.302
MDL Number MFCD00041905
InChI Key GVRNUDCCYWKHMV-UHFFFAOYSA-N
Synonym bis 2-methoxyethyl adipate, hexanedioic acid, bis 2-methoxyethyl ester, bis 2-methoxyethyl hexanedioate, di 2-methoxyethyl adipate, adipic acid, bis 2-methoxyethyl ester, adipic acid bis 2-methoxyethyl ester, hexanedioic acid, 1,6-bis 2-methoxyethyl ester, 1,6-bis 2-methoxyethyl hexanedioate
PubChem CID 66046
IUPAC Name bis(2-methoxyethyl) hexanedioate
SMILES COCCOC(=O)CCCCC(=O)OCCOC

Specifications

Boiling Point 190°C (11 mmHg)
Linear Formula (CH2)4(CO2CH2CH2OCH3)2
Refractive Index 1.4425
Quantity 25 g
Formula Weight 262.3
Percent Purity 98%
Chemical Name or Material Bis (2-methoxyethyl) adipate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.