missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-Hydroxyethyl)-Imino-Tris-(Hydroxymethyl)-Methane, Extra Pure, SLR, Biological Buffer, Fisher Chemical

Product Code. 10704561 Shop All Fisher Chemical Products
Click to view available options
:
100g, Plastic powder jar
This item is not returnable. View return policy

Product Code. 10704561

Brand: Fisher Chemical B/3010/48

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 6976-37-0
Melting Point 103°C
Color White
pH 8.8 to 9.6
Packaging Plastic powder jar
Molecular Formula C8H19NO5
MDL Number 2853
Quantity 100 g
Formula Weight 209.24g/mol
Physical Form Powder
compliance-icons compliance-icons
Product Identifier
  • 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.