Learn More
Diethylene glycol, 99%, extra pure, Thermo Scientific Chemicals
£37.10 - £217.00
Chemical Identifiers
CAS | 111-46-6 |
---|---|
Molecular Formula | C4H10O3 |
Molecular Weight (g/mol) | 106.12 |
MDL Number | MFCD00002882 |
InChI Key | MTHSVFCYNBDYFN-UHFFFAOYSA-N |
Synonym | diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol |
PubChem CID | 8117 |
ChEBI | CHEBI:46807 |
SMILES | OCCOCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10510561
|
Thermo Scientific Acros
121160010 |
1 L | Glass bottle |
£37.10
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10665542
|
Thermo Scientific Acros
121160025 |
2.5 L | Glass bottle |
£71.10
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10649083
|
Thermo Scientific Acros
121160100 |
10 L | Plastic drum |
£217.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-46-6 | |
106.12 | |
MTHSVFCYNBDYFN-UHFFFAOYSA-N | |
8117 | |
OCCOCCO |
C4H10O3 | |
MFCD00002882 | |
diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol | |
CHEBI:46807 |
Specifications
111-46-6 | |
100.0 | |
1.1100g/mL | |
143°C | |
98.5% min. (GC) | |
(HOCH2CH2)2O | |
MFCD00002882 | |
15, 3131 | |
diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol | |
MTHSVFCYNBDYFN-UHFFFAOYSA-N | |
2-(2-hydroxyethoxy)ethan-1-ol | |
8117 | |
38 mPa.s (20°C) | |
99% | |
Oily Liquid |
98.5 | |
-10.0°C | |
245.0°C | |
Authentic | |
C4H10O3 | |
1.4450 to 1.4480 | |
01, 468 | |
1.11 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, ether,, ethylene glycol, insoluble in benzene, toluene, carbon, tetrachloride and aliphatic hydrocarbons | |
OCCOCCO | |
106.12 | |
CHEBI:46807 | |
106.12 | |
Extra Pure | |
Diethylene glycol |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-872-2
- Diethylene glycol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only