missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl)maleate, TRC

Product Code. 17980835
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17980835 100 g 100g
17931875 50 g 50g
17922675 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17980835 Supplier Toronto Research Chemicals Supplier No. TRCB433835100G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-16-5
Molecular Formula C20 H36 O4
Molecular Weight (g/mol) 340.5
Synonym Bis(2-ethylhexyl)maleate, 2-Butenedioic Acid (2Z)-Bis(2-ethylhexyl) Ester, 2-Butenedioic Acid (Z)-Bis(2-ethylhexyl) Ester, Maleic Acid Bis(2-ethylhexyl) Ester, Bis(2-ethylhexyl) Maleate, Ceraphyl 45, DOM, Di-2-ethylhexyl Maleate, Dioctyl Maleate
IUPAC Name bis(2-ethylhexyl) (Z)-but-2-enedioate
SMILES CCCCC(CC)COC(=O)\C=C/C(=O)OCC(CC)CCCC

Specifications

Quantity 100 g
InChI Formula InChI=1S/C20H36O4/c1-5-9-11-17(7-3)15-23-19(21)13-14-20(22)24-16-18(8-4)12-10-6-2/h13-14,17-18H,5-12,15-16H2,1-4H3/b14-13-
Formula Weight 340.2614
Chemical Name or Material Bis(2 ethylhexyl)maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.