Learn More
Bis(2-ethylhexyl) hydrogen phosphate, 95%, Thermo Scientific Chemicals
£199.00 - £531.00
Chemical Identifiers
CAS | 298-07-7 |
---|---|
Molecular Formula | C16H35O4P |
Molecular Weight (g/mol) | 322.43 |
MDL Number | MFCD00009492 |
InChI Key | SEGLCEQVOFDUPX-UHFFFAOYNA-N |
Synonym | bis 2-ethylhexyl hydrogen phosphate, bis 2-ethylhexyl phosphate, hdehp, di 2-ethylhexyl phosphate, dehpa extractant, di 2-ethylhexyl phosphoric acid, escaid 100, d 2ehpa, bis 2-ethylhexyl phosphoric acid |
PubChem CID | 9275 |
IUPAC Name | bis(2-ethylhexyl) hydrogen phosphate |
SMILES | CCCCC(CC)COP(O)(=O)OCC(CC)CCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10075450
|
Thermo Scientific Acros
402470010 |
1 kg | Glass Bottle |
£199.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11925151
|
Thermo Scientific Acros
402470050 |
5 kg | Plastic drum |
£531.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
298-07-7 | |
322.43 | |
SEGLCEQVOFDUPX-UHFFFAOYNA-N | |
9275 | |
CCCCC(CC)COP(O)(=O)OCC(CC)CCCC |
C16H35O4P | |
MFCD00009492 | |
bis 2-ethylhexyl hydrogen phosphate, bis 2-ethylhexyl phosphate, hdehp, di 2-ethylhexyl phosphate, dehpa extractant, di 2-ethylhexyl phosphoric acid, escaid 100, d 2ehpa, bis 2-ethylhexyl phosphoric acid | |
bis(2-ethylhexyl) hydrogen phosphate |
Specifications
298-07-7 | |
100.0 | |
Colorless to Yellow | |
48.0°C (12.0 mmHg) | |
Authentic | |
C16H35O4P | |
1.4400 to 1.4440 | |
01,IV,1786 | |
bis 2-ethylhexyl hydrogen phosphate, bis 2-ethylhexyl phosphate, hdehp, di 2-ethylhexyl phosphate, dehpa extractant, di 2-ethylhexyl phosphoric acid, escaid 100, d 2ehpa, bis 2-ethylhexyl phosphoric acid | |
SEGLCEQVOFDUPX-UHFFFAOYNA-N | |
bis(2-ethylhexyl) hydrogen phosphate | |
9275 | |
95% | |
Bis(2-ethylhexyl) hydrogen phosphate |
94.0 | |
-60.0°C | |
0.9700g/mL | |
137°C | |
95% | |
(CH3(CH2)3CH(C2H5)CH2O)2POOH | |
MFCD00009492 | |
0.97 | |
Solubility in water: slightly soluble | |
CCCCC(CC)COP(O)(=O)OCC(CC)CCCC | |
322.43 | |
322.42 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-056-4
RUO – Research Use Only