missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) adipate, 99%

Product Code. 10547362 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10547362 1 kg Glass bottle 1kg
10167460 250 g Glass bottle 250g
10025353 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10547362 Supplier Thermo Scientific Acros Supplier No. 402460010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-23-1 | C22H42O4 | 370.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-23-1
Molecular Formula C22H42O4
Molecular Weight (g/mol) 370.57
MDL Number MFCD00009496
InChI Key SAOKZLXYCUGLFA-UHFFFAOYSA-N
Synonym bis 2-ethylhexyl adipate, di 2-ethylhexyl adipate, diethylhexyl adipate, deha, plastomoll doa, beha, vestinol oa, bisoflex doa, effomoll doa
PubChem CID 7641
ChEBI CHEBI:34675
IUPAC Name bis(2-ethylhexyl) hexanedioate
SMILES CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -76.0°C
Density 0.9240g/mL
Boiling Point 210.0°C to 218.0°C (5.0 mmHg)
Flash Point 196°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4460 to 1.4480
Quantity 1 kg
Specific Gravity 0.924
Solubility Information Solubility in water: insouble. Other solubilities: soluble in aceton,ether,ethanol
Viscosity 13-15 mPa.s (20°C)
Formula Weight 370.57
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) adipate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.