missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) adipate, 99%

Product Code. 11348295
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348295 100 g 100g
11338295 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348295 Supplier Thermo Scientific Alfa Aesar Supplier No. L10195.22
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 103-23-1 | C22H42O4 | 370.574 g/mol

It is employed as a plasticizer. It is a skin conditioning solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a plasticizer. It is a skin conditioning solvent.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-23-1
Molecular Formula C22H42O4
Molecular Weight (g/mol) 370.574
MDL Number MFCD00009496
InChI Key SAOKZLXYCUGLFA-UHFFFAOYSA-N
Synonym bis 2-ethylhexyl adipate, di 2-ethylhexyl adipate, diethylhexyl adipate, deha, plastomoll doa, beha, vestinol oa, bisoflex doa, effomoll doa
PubChem CID 7641
ChEBI CHEBI:34675
IUPAC Name bis(2-ethylhexyl) hexanedioate
SMILES CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC

Specifications

Melting Point -67°C
Density 0.99
Boiling Point 166°C to 168°C (1 mmHg)
Flash Point 196°C (384°F)
Refractive Index 1.447
Quantity 100 g
Beilstein 1803774
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 370.58
Percent Purity 99%
Chemical Name or Material Bis(2-ethylhexyl) adipate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.