missing translation for 'onlineSavingsMsg'
Learn More

Bis[2-(diphenylphosphino)ethyl]ammonium chloride, 97+%

Product Code. 15455596
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455596 250 mg 250mg
15435596 1 g 1g
15445596 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15455596 Supplier Thermo Scientific Alfa Aesar Supplier No. H60250.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 66534-97-2 | C28H30ClNP2 | 477.95 g/mol

It is involved in the Synthesis of the new chiral aminodiphosphine ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is involved in the Synthesis of the new chiral aminodiphosphine ligands.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66534-97-2
Molecular Formula C28H30ClNP2
Molecular Weight (g/mol) 477.95
MDL Number MFCD00061569
InChI Key AVDUVHMGGDOIQI-UHFFFAOYSA-N
Synonym bis 2-diphenylphosphino ethyl amine hydrochloride, bis 2-diphenylphosphino ethyl ammonium chloride, bis 2-diphenylphosphanyl ethyl amine hydrochloride, ethanamine, 2-diphenylphosphino-n-2-diphenylphosphino ethyl-, hydrochloride, 2-diphenylphosphanyl-n-2-diphenylphosphanyl ethyl ethan-1-amine-hydrogen chloride 1/1
PubChem CID 21144848
IUPAC Name 2-diphenylphosphanyl-N-(2-diphenylphosphanylethyl)ethanamine;hydrochloride
SMILES Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)NCCP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 175°C to 180°C
Linear Formula [(C6H5)2PCH2CH2]2NH2Cl
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 477.95
Percent Purity ≥97%
Chemical Name or Material Bis[2-(diphenylphosphino)ethyl]ammonium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.