missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-(dimethylamino)ethyl) Succinate, TRC

Product Code. 30132826
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132826 50 mg 50mg
17934825 100 mg 100mg
17923575 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132826 Supplier Toronto Research Chemicals Supplier No. TRCB40022350MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19249-04-8
Molecular Formula C12 H24 N2 O4
Molecular Weight (g/mol) 260.33
Synonym Butanedioic Acid Bis[2-(dimethylamino)ethyl] Ester
IUPAC Name bis[2-(dimethylamino)ethyl] butanedioate
SMILES CN(C)CCOC(=O)CCC(=O)OCCN(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H24N2O4/c1-13(2)7-9-17-11(15)5-6-12(16)18-10-8-14(3)4/h5-10H2,1-4H3
Formula Weight 260.1736
Chemical Name or Material Bis(2-(dimethylamino)ethyl) Succinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.