missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) selenide, 99.999% (metals basis)

Product Code. 11332228
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332228 5 g 5g
11322228 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332228 Supplier Thermo Scientific Alfa Aesar Supplier No. 013130.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1315-05-5

It is used to prepare large zintl ion, it is now actively explored for thin film solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1315-05-5
MDL Number MFCD00016319

Specifications

Melting Point 611°C
Physical Form Pieces
Quantity 5 g
UN Number UN3077
Merck Index 14,712
Solubility Information Very slightly soluble in water
Formula Weight 480.38
Percent Purity 99.999%
Sensitivity Moisture sensitive
Assay (metals basis)
Chemical Name or Material Antimony(III) selenide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.