missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-[di(1-adamantyl)phosphino]ethyl)amine, 97+%

Product Code. p-7040939
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436056 250 mg 250mg
15417358 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436056 Supplier Thermo Scientific Alfa Aesar Supplier No. H60853.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1086138-36-4 | C44H69NP2 | 673.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1086138-36-4
Molecular Formula C44H69NP2
Molecular Weight (g/mol) 673.99
MDL Number MFCD17018779
InChI Key KBHQOEPPHGCYHC-UHFFFAOYSA-N
Synonym bis 2-di adamantan-1-yl phosphino ethyl amine, bis 2-di-1-adamantylphosphino ethyl amine, bis 2-bis adamantan-1-yl phosphanyl ethyl amine, bis 2-diadamantylphosphino ethyl amine, bis 2-di 1-adamantyl phosphino ethyl amine, bis 2-di-1-adamantylphosphino ethyl amine,min, bis 2-bis 3r,5s,7s-adamantan-1-yl phosphanyl ethyl amine, 2-di tricyclo 3.3.1.1∼3,7∼ decan-1-yl phosphanyl-n-2-di tricyclo 3.3.1.1∼3,7∼ decan-1-yl phosphanyl ethyl ethan-1-amine
PubChem CID 58194772
IUPAC Name 2-[bis(1-adamantyl)phosphanyl]-N-[2-[bis(1-adamantyl)phosphanyl]ethyl]ethanamine
SMILES C(CP(C12CC3CC(CC(C3)C1)C2)C12CC3CC(CC(C3)C1)C2)NCCP(C12CC3CC(CC(C3)C1)C2)C12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 217°C to 222°C
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 673.97
Percent Purity ≥97%
Chemical Name or Material Bis(2-[di(1-adamantyl)phosphino]ethyl)amine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.