missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-chloroethyl) ether, 99%

Product Code. 11482347
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482347 250 g 250g
11472347 50 g 50g
11462347 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482347 Supplier Thermo Scientific Alfa Aesar Supplier No. A14234.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-44-4 | C4H8Cl2O | 143.01 g/mol

Bis(2-chloroethyl) ether is used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-chloroethyl) ether is used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.

Solubility
Soluble in water. (1.1 g/L) at 20°C.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-44-4
Molecular Formula C4H8Cl2O
Molecular Weight (g/mol) 143.01
MDL Number MFCD00000975
InChI Key ZNSMNVMLTJELDZ-UHFFFAOYSA-N
Synonym 2-chloroethyl ether, 2,2'-dichlorodiethyl ether, bis 2-chloroethyl ether, chlorex, chloroethyl ether, dcee, dichloroether, khloreks, dichloroethyl ether
PubChem CID 8115
ChEBI CHEBI:34573
IUPAC Name 1-chloro-2-(2-chloroethoxy)ethane
SMILES ClCCOCCCl

Specifications

Melting Point -50°C
Density 1.219
Boiling Point 178°C
Flash Point 55°C (131°F)
Odor Pungent
Linear Formula ClCH2CH2OCH2CH2Cl
Refractive Index 1.457
Quantity 250 g
UN Number UN1916
Beilstein 605317
Merck Index 14,3066
Solubility Information Soluble in water. (1.1g/L) at 20°C.
Formula Weight 143.01
Percent Purity 99%
Chemical Name or Material Bis(2-chloroethyl) ether
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bis(2-chloroethyl) ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Carcinogenicity Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contain: Bis(2-chloroethyl) ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.