missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-chloroethyl) Ether(1,1'-Oxybis[2-chloro-ethane]), TRC

Product Code. 30423691
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30423691

Brand: Toronto Research Chemicals TRCO8705255G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis-(2-chloroethyl)ether
CAS 111-44-4
Molecular Formula C4 H8 Cl2 O
Quantity 5 g
Synonym Ether','bis(2-chloroethyl) (8CI), Ether','bis(chloroethyl) (7CI), 1,1'-Oxybis[2-chloroethane], 1,5-Dichloro-3-oxapentane, 1-Chloro-2-(2-chloroethoxy)ethane, 1-Chloro-2-(β-chloroethoxy)ethane, 2,2'-Dichlorethyl ether, 2,2'-Dichlorodiethyl ether, 2,2'-Dichloroethyl ether, 2-(2-Chloroethoxy)ethyl chloride, 2-Chloroethyl ether, BCEE, Bis(2-chloroethyl) ether, Bis(β-chloroethyl) ether, Chlorex, DCEE, Di(β-Chloroethyl) ether, Di-2-chloroethyl ether, Diethylene glycol dichloride, NSC 406647, Oxygen mustard, sym-Dichloroethyl ether, β,β'-Dichlorodiethyl ether, β,β'-Dichloroethyl ether
InChI Formula InChI=1S/C4H8Cl2O/c5-1-3-7-4-2-6/h1-4H2
SMILES ClCCOCCCl
IUPAC Name 1-chloro-2-(2-chloroethoxy)ethane
Molecular Weight (g/mol) 143.01
Formula Weight 141.9952
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.