missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-bromoethyl) ether, 95%

Product Code. 11406780
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406780 5 g 5g
11416780 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11406780 Supplier Thermo Scientific Alfa Aesar Supplier No. L11510.06
  • ONLINE EXCLUSIVE PRICE
    £40.90
    £36.65 / 5g
    Save £4.25
    10% Off
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5414-19-7 | C4H8Br2O | 231.915 g/mol

Bis(2-bromoethyl) ether is used dfor synthesis of heterocyclic and crown ether. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-bromoethyl) ether is used dfor synthesis of heterocyclic and crown ether. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5414-19-7
Molecular Formula C4H8Br2O
Molecular Weight (g/mol) 231.915
MDL Number MFCD00039196
InChI Key FOZVXADQAHVUSV-UHFFFAOYSA-N
Synonym bis 2-bromoethyl ether, 1-bromo-2-2-bromoethoxy ethane, 2-bromoethyl ether, 2,2'-dibromodiethyl ether, ethane, 1,1'-oxybis 2-bromo, ether, bis 2-bromoethyl, beta,beta'-dibromodiethyl ether, 2,2-dibromodiethyl ether, 2-bromoethylether
PubChem CID 21521
IUPAC Name 1-bromo-2-(2-bromoethoxy)ethane
SMILES C(CBr)OCCBr

Specifications

Density 1.845
Boiling Point 92°C to 93°C (12 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.514
Quantity 5 g
UN Number UN2810
Beilstein 1098335
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 231.93
Percent Purity 96%
Chemical Name or Material Bis(2-bromoethyl) ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.