missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-aminophenyl) ether, 98%, Thermo Scientific Chemicals

Product Code. 15481618
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is catalytic reagent and used in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is catalytic reagent and used in pharmaceutical industry.

Solubility
Slightly soluble in water.

Notes
Store at low temperature, away from light and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24878-25-9
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.241
MDL Number MFCD00792533
InChI Key GOJFAKBEASOYNM-UHFFFAOYSA-N
Synonym 2,2'-oxydianiline, 2-2-aminophenoxy aniline, 2,2 inverted exclamation marka-oxydianiline, aminophenyl ether, o,o'-oxydianiline, 2-aminophenyl ether, bis 2-aminophenyl ether, 2,2'-oxybis benzenamine, benzenamine,2,2'-oxybis, 2,2'-diaminodiphenyl ether
PubChem CID 458824
IUPAC Name 2-(2-aminophenoxy)aniline
SMILES C1=CC=C(C(=C1)N)OC2=CC=CC=C2N

Specifications

Melting Point 61°C to 64°C
Color Brown to White
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 200.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Bis(2-aminophenyl) ether

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'