missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-aminoethyl)(nitroso)amine Dihydrochloride, TRC

Product Code. 30382225
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382225 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382225 Supplier Toronto Research Chemicals Supplier No. TRCE74165510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(2-aminoethyl)(nitroso)amine Dihydrochloride
CAS 239096-66-3
Molecular Formula C4H12N4O • 2HCl
Quantity 10 mg
Synonym N-(2-Aminoethyl)-N-nitroso-1,2-ethanediamine Dihydrochloride
InChI Formula InChI=1S/C4H12N4O.2ClH/c5-1-3-8(7-9)4-2-6;;/h1-6H2;2*1H
SMILES NCCN(CCN)N=O.Cl.Cl
IUPAC Name N,N-bis(2-aminoethyl)nitrous amide hydrogen chloride
Molecular Weight (g/mol) 205.08
Formula Weight 204.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.